missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Azide, 0.5% (w/v), Ricca Chemical
APHA for Chromium Testing
Supplier: Ricca Chemical Company 714416
Specifications
| 500 mL | |
| 500 mL | |
| Sodium Azide |
| Natural Poly Bottle | |
| APHA for Chromium |