missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ascorbate, USP, bioCERTIFIED™, 1 kg, Spectrum Chemical

Catalog No. 18615351
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18615351 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18615351 Supplier Spectrum Chemical Mfg Cor Supplier No. SO9051KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-quality USP grade chemical manufactured under cGMP for biopharmaceutical applications.

bioCERTIFIED™ products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Spectrum Chemical's bioCERTIFIED™ quality management program goes beyond standard testing and includes APIs, excipients and many more chemicals that carry additional certifications and documentation specifically designed to meet the needs of biopharmaceutical businesses including manufacturers and pharmacies. Each bioCERTIFIED™ chemical undergoes expanded testing for bioburden, endotoxin, elemental impurities and other tests required by the monographs.

Chemical Identifiers

CAS 134-03-2
Molecular Weight (g/mol) 198.11

Specifications

Quantity 1 kg
Grade USP
Physical Form Crystalline Powder
Chemical Name or Material Sodium Ascorbate