missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Ascorbate, Powder, USP, 99-101%, Spectrum™ Chemical
Description
Spectrum™ Chemical Sodium Ascorbate, Powder, USP is used to prevent or treat low levels of vitamin C in people who do not get enough of the vitamin from their diets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| pH | 7.0 to 8.0 |
| Loss on Drying | 0.0025 |
| Packaging | Fiber Drum |
| Quantity | 50 kg |
| Specific Rotation | +103° to +108° |
| Percent Purity | 99 to 101% |
| Grade | USP |
| Physical Form | Powder |
| Chemical Name or Material | Sodium Ascorbate |