missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ascorbate, Powder, USP, 99-101%, Spectrum™ Chemical

Catalog No. 18606307
Change view
Click to view available options
Quantity:
5 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606307 5 kg Poly Pail
18606308 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18606307 Supplier Spectrum Chemical Mfg Cor Supplier No. S13495KG
Get Up to $500 in Chemicals with Purchase
View all promotions

S1349, 134-03-2, C6H7NaO6

Spectrum™ Chemical Sodium Ascorbate, Powder, USP is used to prevent or treat low levels of vitamin C in people who do not get enough of the vitamin from their diets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

CAS Max % 1
pH 7.0 to 8.0
Loss on Drying 0.0025
Packaging Poly Pail
Quantity 5 kg
Specific Rotation +103° to +108°
Percent Purity 99 to 101%
Grade USP