missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ascorbate, Powder, USP, 99-101%, Spectrum™ Chemical

Catalog No. 18606305
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606305 100 g Amber Glass Bottle
18606306 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18606305 Supplier Spectrum Chemical Mfg Cor Supplier No. S1349100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

S1349, 134-03-2, C6H7NaO6

Spectrum™ Chemical Sodium Ascorbate, Powder, USP is used to prevent or treat low levels of vitamin C in people who do not get enough of the vitamin from their diets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

CAS Max % 1
pH 7.0 to 8.0
Loss on Drying 0.0025
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation +103° to +108°
Percent Purity 99 to 101%
Grade USP