missing translation for 'onlineSavingsMsg'
Learn More

Sodium Ascorbate, FCC, 99-101%, Spectrum™ Chemical

Catalog No. p-7158179
Change view
Click to view available options
Quantity:
500 g
5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606302 500 g Amber Glass Bottle
18606303 5 kg Poly Pail
18606304 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606302 Supplier Spectrum Chemical Mfg Cor Supplier No. S1137500GM
Only null left

S1137, 134-03-2, C6H7NaO6

Spectrum™ Chemical Sodium Ascorbate, FCC is used as a food additive as an antioxidant and acidity regulator. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

CAS Max % 1
Loss on Drying 0.0025
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 99 to 101%
Grade FCC