missing translation for 'onlineSavingsMsg'
Learn More

Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade

Catalog No. p-4407859
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4032022 100 g
AA4032036 500 g
AA40320A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4032022 Supplier Thermo Scientific Chemicals Supplier No. 04032022
Only null left

CAS: 7783-13-3 | H13NNaO8P | 209.067 g/mol

Used in the laboratory as an essential ingredient of the microcosmic salt bead test for identification of metallic radicals. Also used as intermediates. Finds application in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7783-13-3
Molecular Formula H13NNaO8P
Molecular Weight (g/mol) 209.067
MDL Number MFCD00066970
InChI Key IQTQISLCLWJRPM-UHFFFAOYSA-M
Synonym unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate
PubChem CID 56924540
IUPAC Name azanium;sodium;hydrogen phosphate;tetrahydrate
SMILES [NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+]

Specifications

Melting Point 79°C (decomposition)
Color White
Physical Form Crystals
Quantity 100 g
Solubility Information Soluble in water. Insoluble in alcohol
Formula Weight 209.07 (137.02 Anhydrous)
Grade Reagent
Odor Odorless
Appearance White crystals
Density 1.544
Chemical Name or Material Sodium ammonium hydrogen phosphate tetrahydrate
Show More Show Less
Hazard Category H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only