missing translation for 'onlineSavingsMsg'
Learn More

Sodium amide, 95%, pure

Catalog No. p-4509790
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC197051000 100 g Glass bottle
AC197055000 500 g Glass bottle
AC197050025 2.5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC197051000 Supplier Thermo Scientific Chemicals Supplier No. 197051000
Only null left

CAS: 7782-92-5 | H2NNa | 39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7782-92-5
InChI Key ODZPKZBBUMBTMG-UHFFFAOYSA-N
Synonym sodium amide, sodamide, sodium amide na nh2, unii-5db3g6px9d, 5db3g6px9d, sodium azanide, sodium amide, 50 wt. % suspension in toluene, sodiumamide, natriumamid, sodioamine
PubChem CID 24533
IUPAC Name sodium;azanide
SMILES [NH2-].[Na+]

Specifications

Boiling Point 400°C
Melting Point 210°C
Color Gray-White to White
Physical Form Powder, Irregular Flakes or Granules
Quantity 100 g
Assay Percent Range 95%
Molecular Formula H2NNa
Linear Formula NaNH2
Fieser 01,1034; 02,373; 04,439; 05,591; 06,525; 08,446; 15,288
Merck Index 15,8714
Solubility Information Solubility in water: reacts violently with water. Other solubilities: decomposes in hot alcohol
Molecular Weight (g/mol) 39
Formula Weight 39
Percent Purity 95%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Sodium amide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Very toxic to aquatic life.
Reacts violently with water.
May form explosive peroxides.<
Precautionary Statement GHS P Statement
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present
EINECSNumber 231-971-

RUO – Research Use Only