Learn More
Sodium aluminum oxide, tech.
CAS: 1302-42-7 | Al2O3·Na2O
$127.47 - $224.19
Chemical Identifiers
| CAS | 1302-42-7 |
|---|---|
| Molecular Formula | Al2O3·Na2O |
| MDL Number | MFCD00801010 |
| Synonym | Sodium aluminate |
Description
Sodium aluminum oxide is used in the isomerization of alkene and isomerization of amines. It is also used in water treatment as an adjunct and as a coagulant aid to flocculation. It is involved in the removal of silica and phosphates from waste water. It is employed to accelerate concrete solidification. It finds application in the paper industry and fire brick production. It acts as an intermediate in the production of zeolites.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1302-42-7 | |
| Al2O3·Na2O | |
| MFCD00801010 | |
| Sodium aluminate |
Specifications
| White | |
| Crystalline | |
| Al2O3·Na2O | |
| Sodium aluminate | |
| 163.94 | |
| White crystalline | |
| Sodium aluminum oxide |
| 1302-42-7 | |
| 100 g | |
| MFCD00801010 | |
| Insoluble in alcohol,soluble in waterInsoluble in alcohol. Soluble in water. | |
| Hygroscopic | |
| 1.52 |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : DOT Class: 8; Packing Group: III
EINECSNumber : 215-100-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only