missing translation for 'onlineSavingsMsg'
Learn More

Sodium Alginate, NF, 90.8-106%, Spectrum™ Chemical

Catalog No. 18606287
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18606287 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606287 Supplier Spectrum Chemical Mfg Cor Supplier No. S1118500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1118, 9005-38-3, (C6H7O6Na)n

Spectrum™ Chemical Sodium Alginate, NF is a gum that has no discernible flavor and is used in the food and pharmaceutical industries as an emulsifier and to increase viscosity. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
SMILES [Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O

Specifications

CAS Max % 1
Physical Form Flakes or Powder
Quantity 500 g
Percent Purity 90.8 to 106%
Grade NF
Loss on Drying 0.15
Packaging Poly Bottle
Chemical Name or Material Sodium Alginate