missing translation for 'onlineSavingsMsg'
Learn More

Sodium Alginate, NF, 90.8-106%, Spectrum™ Chemical

Catalog No. 18606288
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606288 2.5 kg Poly Pail
18606289 12 kg Poly Pail
18606290 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606288 Supplier Spectrum Chemical Mfg Cor Supplier No. S11182.5KG

S1118, 9005-38-3, (C6H7O6Na)n

Spectrum™ Chemical Sodium Alginate, NF is a gum that has no discernible flavor and is used in the food and pharmaceutical industries as an emulsifier and to increase viscosity. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
SMILES [Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 90.8 to 106%
Grade NF
Loss on Drying 0.15
Packaging Poly Pail