missing translation for 'onlineSavingsMsg'
Learn More

Sodium Alginate, MP Biomedicals

Catalog No. ICN218295
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218295 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218295 Supplier MP Biomedicals Inc Supplier No. 0521829580

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa

Specifications

Physical Form Powder
Quantity 100 g
Chemical Name or Material Sodium Alginate