missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate, Trihydrate, Granular, USP, 99-101%, Spectrum™ Chemical
Description
Spectrum™ Chemical Sodium Acetate, Trihydrate, Granular, USP is used as a source of sodium ions in solutions for dialysis and as a systemic and urinary alkalizer, diuretic, and expectorant. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| pH | 7.5 9.2 |
| CAS Max % | 1 |
| Percent Purity | 99 to 101% |
| Grade | USP |
| Quantity | 500 g |
| Loss on Drying | 38-41% |
| Packaging | Poly Bottle |