missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Trihydrate, Granular, USP, 99-101%, Spectrum™ Chemical

SO105, 6131-90-4, NaC2H3O2·3H2O

Supplier:  Spectrum Chemical Mfg Cor SO10550KGBL

 Afficher plus de versions de ce produit

Numéro de catalogue. 18606276


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spectrum™ Chemical Sodium Acetate, Trihydrate, Granular, USP is used as a source of sodium ions in solutions for dialysis and as a systemic and urinary alkalizer, diuretic, and expectorant. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Spécifications

Spécifications

7.5 9.2
1
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium acetate trihydrate
99 to 101%
50 kg
Fiber Drum
6131-90-4
C2H9NaO5
O.O.O.[Na+].CC([O-])=O
136.08
USP
38-41%

Identifiants chimiques

6131-90-4
136.08
sodium acetate trihydrate
C2H9NaO5
AYRVGWHSXIMRAB-UHFFFAOYSA-M
O.O.O.[Na+].CC([O-])=O
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions