missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Trihydrate, Granular, USP, 99-101%, Spectrum™ Chemical

Catalog No. 18606274
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
100 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606274 2.5 kg Poly Bottle
18606275 12 kg Poly Pail
18606276 50 kg Fiber Drum
18606277 100 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606274 Supplier Spectrum Chemical Mfg Cor Supplier No. SO1052.5KG

SO105, 6131-90-4, NaC2H3O2·3H2O

Spectrum™ Chemical Sodium Acetate, Trihydrate, Granular, USP is used as a source of sodium ions in solutions for dialysis and as a systemic and urinary alkalizer, diuretic, and expectorant. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.08
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
IUPAC Name sodium acetate trihydrate
SMILES O.O.O.[Na+].CC([O-])=O

Specifications

pH 7.5 9.2
CAS Max % 1
Percent Purity 99 to 101%
Grade USP
Quantity 2.5 kg
Loss on Drying 38-41%
Packaging Poly Bottle