missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate, Trihydrate, Crystal, ACS, 99.0-101.0%, Spectrum™ Chemical
Specifications
Specifications
| pH | 7.5 to 9.2 |
| CAS Max % | 1 |
| Quantity | 500 g |
| Percent Purity | 99 to 101% |
| Grade | ACS |
| Packaging | Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.