missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Trihydrate, Crystal, ACS, 99.0-101.0%, Spectrum™ Chemical

Catalog No. 18613248
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18613248 500 g Poly Bottle
18613249 2.5 kg Poly Bottle
18613250 12 kg Poly Pail
18613251 50 kg Fiber Drum
4 options

Catalog No. 18613248

Supplier: Spectrum Chemical Mfg Cor S1115500GM

Only null left

Chemical Identifiers

CAS 6131-90-4

Specifications

pH 7.5 to 9.2
CAS Max % 1
Quantity 500 g
Percent Purity 99 to 101%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.