missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate trihydrate, ACS, 99.0%-101%

Catalog No. p-6827150
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1155336 500 g
AA11553A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1155336 Supplier Thermo Scientific Chemicals Supplier No. 01155336
Only null left

CAS: 6131-90-4 | C2H9NaO5 | 136.079 g/mol

In photography, analytical laboratory as buffer and in pharmaceutical.Sodium acetate trihydrate is used in various applications. It is used as a buffer of a pH range in between to 4.0 to 6.0. It is also used in the purification and precipitation of nucleic acids and in protein crystallization. It is used as a mordant dyeing, as a pickling agent in chrome tanning, as a neutralizer in waste streams containing sulfuric acid, in the textile industry and as a photoresist while using aniline dyes. It plays an important role to retard vulcanization of chloroprene in synthetic rubber production. It is a precursor to prepare ester from alkyl halide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
MDL Number MFCD00071557
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Melting Point 62°C to 64°C
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.0% to 100.5%
Beilstein 3732037
Solubility Information Freely soluble in alcohol. Soluble in water and alcohol.
Formula Weight 136.08 (82.03 Anhydrous)
Grade ACS Reagent
Density 1.45
Chemical Name or Material Sodium Acetate Trihydrate, For ACS analysis
Hazard Category H500
EINECSNumber 204-823-8
RTECSNumber AJ4580000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only