missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate trihydrate, 99%

Catalog No. p-7023140
Change view
Click to view available options
Quantity:
10,000 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1623036 500 g
AAA162300E 2500 g
AAA162300C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1623036 Supplier Thermo Scientific Chemicals Supplier No. A1623036
Only null left

CAS: 6131-90-4 | C2H9NaO5 | 136.079 g/mol

Sodium acetate trihydrate is used in various applications. It is used as a buffer of pH range in between to 4.0 to 6.0. It is also used in the purification and precipitation of nucleic acids and in protein crystallization. It is used as mordant dyeing, as a pickling agent in chrome tanning, as a neutralizer in waste streams containing sulfuric acid in textile industry and as a photoresist while using aniline dyes. It plays an important role to retard vulcanization of chloroprene in synthetic rubber production. It is a precursor to prepare ester from alkyl halide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium acetate trihydrate is used in various applications. It is used as a buffer of pH range in between to 4.0 to 6.0. It is also used in the purification and precipitation of nucleic acids and in protein crystallization. It is used as mordant dyeing, as a pickling agent in chrome tanning, as a neutralizer in waste streams containing sulfuric acid in textile industry and as a photoresist while using aniline dyes. It plays an important role to retard vulcanization of chloroprene in synthetic rubber production. It is a precursor to prepare ester from alkyl halide.

Solubility
Soluble in water and alcohol.

Notes
-3H{2}O @ 120°. Incompatible with strong oxidizing agents and halogens.

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
MDL Number MFCD00071557
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Melting Point 62°C to 64°C
Density 1.45
pH 7.5 to 9.0
Odor Odorless
Quantity 500 g
Beilstein 3732037
Merck Index 14,8571
Solubility Information Soluble in water and alcohol.
Formula Weight 136.08 (82.03 Anhydrous)
Percent Purity 99%
Chemical Name or Material Sodium acetate trihydrate
Show More Show Less
Hazard Category H500
EINECSNumber 204-823-8
RTECSNumber AJ4580000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only