missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, TraceSELECT™, Honeywell Fluka™

Catalog No. p-7072219
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002757 25 g
6002756 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002757 Supplier Honeywell Chemicals Supplier No. 5992925G
Only null left

TraceSELECT™, for trace analysis,≥99.999% (metals basis), anhydrous

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium;acetate
SMILES [Na+].CC([O-])=O

Specifications

Quantity 25 g
Linear Formula CH3COONa
UN Number NONH for all modes of transport
Beilstein 3595639
Flash Point <250°C (>482°F)
Formula Weight 82.03g/mol
Percent Purity ≥99.99% (metals basis)
Grade TraceSELECT
Ignition Point 1112°F
Chemical Name or Material Sodium acetate
RTECSNumber AJ4300010