missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, MP Biomedicals™

Catalog No. MP21954965
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21954965 500 g
MP219549602 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP21954965 Supplier MP Biomedicals Inc Supplier No. 02195496.5
Only null left

Sodium acetate Anhydrous, ACS, White Crystalline Powder

  • It is used as a buffer in the buffering range of pH 3.6-5.6.
  • It is also used in the purification and precipitation of nucleic acids, protein crystallization, Staining of gels in protein gel electrophoresis, HPLC, mordant dyeing, DNA microarray studies of E. coli response, investigation of protein unfolding during reverse phase chromatography.
  • It is used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents.
  • It is also used as substrate for acetokinase. Sodium acetate is the sodium salt of acetic acid.

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym Acetic acid sodium salt, Anhydrous Sodium Acetate
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

Boiling Point >400°C with decomposition (literature)
Melting Point 324°C with decomposition (literature)
Color White
pH 7.0 to 9.2 (5% aq soln)
Physical Form Crystalline Powder
Quantity 500 g
Beilstein 3595639
Flash Point >250°C / >482°F (lit.)
Solubility Information Soluble freely in water; moderately soluble in alcohol.
Grade ACS
Loss on Drying ≤1.0% (120°C)
Vapor Pressure ca. 0 hPa at 20°C(lit.)
Density 1.45g/cm3(lit.)
Assay ≥99.0%
Show More Show Less
RTECSNumber AJ4300010
storageNote1 Store at Room Temperature (15-30°C), dessicate.