missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate, For HPLC, ≥99.0%, Honeywell Fluka™
for HPLC,≥99.0%
$1043.53 - $1043.53
Chemical Identifiers
| CAS | 127-09-3 |
|---|---|
| Molecular Formula | C2H3NaO2 |
| Molecular Weight (g/mol) | 82.03 |
| MDL Number | MFCD00012459 |
| InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
| Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
| PubChem CID | 517045 |
| ChEBI | CHEBI:32954 |
| IUPAC Name | sodium;acetate |
| SMILES | [Na+].CC([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 127-09-3 | |
| 82.03 | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| 517045 | |
| sodium;acetate |
| C2H3NaO2 | |
| MFCD00012459 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| CHEBI:32954 | |
| [Na+].CC([O-])=O |
Specifications
| 127-09-3 | |
| C2H3NaO2 | |
| MFCD00012459 | |
| 3595639 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| [Na+].CC([O-])=O | |
| 82.03 | |
| CHEBI:32954 | |
| ≥99% (NT) | |
| Glass bottle |
| 250 g | |
| CH3COONa | |
| NONH for all modes of transport | |
| <250°C (>482°F) | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| sodium;acetate | |
| 517045 | |
| 82.03g/mol | |
| 1112°F | |
| Sodium acetate |
Safety and Handling
RTECSNumber : AJ4300010