missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Anhydrous, USP, 99-101%, Spectrum™ Chemical

Catalog No. 18606268
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606268 50 kg Fiber Drum
18606266 2.5 kg Poly Pail
18606267 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606268 Supplier Spectrum Chemical Mfg Cor Supplier No. SO10450KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SO104, 127-09-3, C2H3NaO2

Spectrum™ Chemical Sodium Acetate, Anhydrous, USP is used as an electrolyte replenisher in isosmotic solution for parenteral replacement of acute losses of extracellular fluid without disturbing normal electrolyte balance. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

 

Note: This item is non-returnable.

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

pH 7.5 to 9.2
CAS Max % 1
Quantity 50 kg
Loss on Drying 0.01
Packaging Fiber Drum
Percent Purity 99 to 101%
Grade USP