missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium Acetate, Anhydrous, OmniPur™, Calbiochem™,

Catalog No. 7510500GM
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7510500GM 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 7510500GM Supplier MilliporeSigma™ Supplier No. 7510500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Molecular biology grade sodium acetate anhydrous for laboratory and manufacturing applications.

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

Physical Form Crystals
Quantity 500 g
Assay Percent Range 99% min
Formula Weight 82.03
Percent Purity ≥99.0%
Grade Molecular Biology
Chemical Name or Material Sodium Acetate, Anhydrous
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.