missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Anhydrous, FCC, 99-101%, Spectrum™ Chemical

Catalog No. 18606262
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606262 2.5 kg Poly Pail
18606263 12 kg Poly Pail
18606264 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606262 Supplier Spectrum Chemical Mfg Cor Supplier No. S11122.5KG

S1112, 127-09-3, C2H3NaO2

Spectrum™ Chemical Sodium Acetate, Anhydrous, FCC is used for controlling the pH level of food items and acts as a preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

CAS Max % 1
Percent Purity 99 to 101%
Grade FCC
Quantity 2.5 kg
Loss on Drying 0.01
Packaging Poly Pail