missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$5418.67 - $22628.22
Chemical Identifiers
| CAS | 127-09-3 |
|---|---|
| Molecular Formula | C2H3NaO2 |
| Molecular Weight (g/mol) | 82.03 |
| MDL Number | MFCD00012459 |
| InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
| Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
| PubChem CID | 517045 |
| ChEBI | CHEBI:32954 |
| SMILES | [Na+].CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
02003978
|
Avantor J.T.Baker
347001 |
500 g | Poly Bottle |
Case of 4 Each for $5,418.67
|
|
||||
|
02003975
|
Avantor J.T.Baker
347005 |
2.5 kg | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 127-09-3 | |
| 82.03 | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| 517045 | |
| [Na+].CC([O-])=O |
| C2H3NaO2 | |
| MFCD00012459 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| CHEBI:32954 |
Specifications
| 127-09-3 | |
| C2H3NaO2 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| [Na+].CC([O-])=O | |
| 82.03 | |
| CHEBI:32954 | |
| BAKER ANALYZED™ ACS Reagent | |
| 1L = 1.53kg |
| 500 g | |
| MFCD00012459 | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| sodium;acetate | |
| 517045 | |
| 82.03 | |
| Poly Bottle | |
| Sodium Acetate, Anhydrous |