missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Anhydrous, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7166278
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18613244 125 g Poly Bottle
18613246 500 g Poly Bottle
18613239 2.5 kg Poly Bottle
18613245 12 kg Poly Pail
18613247 50 kg Fiber Drum
5 options

Catalog No. 18613244

Supplier: Spectrum Chemical Mfg Cor S1110125GM

Only null left

Chemical Identifiers

CAS 127-09-3

Specifications

pH 7.0 to 9.2
CAS Max % 1
Quantity 125 g
Percent Purity 99%
Grade ACS
Identification To Pass Test
Loss on Drying 0.01
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.