Learn More
Sodium acetate, anhydrous, ACS, 99.0% min
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
$76.63 - $729.07
Chemical Identifiers
| CAS | 127-09-3 |
|---|---|
| Molecular Formula | C2H3NaO2 |
| Molecular Weight (g/mol) | 82.03 |
| MDL Number | MFCD00012459 |
| InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
| Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
| PubChem CID | 517045 |
| ChEBI | CHEBI:32954 |
| IUPAC Name | sodium acetate |
| SMILES | [Na+].CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1155430
|
Thermo Scientific Chemicals
01155430 |
250 g |
Each for $76.63
|
|
|||||
|
AA11554A1
|
Thermo Scientific Chemicals
011554A1 |
1 kg |
Each for $208.30
|
|
|||||
|
AA11554A7
|
Thermo Scientific Chemicals
011554A7 |
5 kg |
Each for $729.07
|
|
|||||
Description
Sodium acetate is usually used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography and it is also a plating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 127-09-3 | |
| 82.03 | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| 517045 | |
| sodium acetate |
| C2H3NaO2 | |
| MFCD00012459 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| CHEBI:32954 | |
| [Na+].CC([O-])=O |
Specifications
| 324°C | |
| White | |
| 250 g | |
| C2H3NaO2 | |
| 3595639 | |
| Soluble in water,ethanol,methanol and hydrazine. | |
| [Na+].CC([O-])=O | |
| 82.03 | |
| CHEBI:32954 | |
| ACS | |
| Hygroscopic | |
| 1.528 g/cm3 at 20°C |
| 127-09-3 | |
| Crystalline | |
| 99.0% min. | |
| MFCD00012459 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| sodium acetate | |
| 517045 | |
| 82.03 | |
| Nearly odorless | |
| White crystalline powder | |
| Sodium acetate, anhydrous, For ACS analysis |
Safety and Handling
H500
EINECSNumber : 204-823-8
RTECSNumber : AJ4300010
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only