missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, anhydrous, ACS, 99.0% min

Catalog No. AA11554A7
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11554A7 5 kg
AA1155430 250 g
AA11554A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA11554A7 Supplier Thermo Scientific Chemicals Supplier No. 011554A7

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

Sodium acetate is usually used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography and it is also a plating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 324°C
Color White
Physical Form Crystalline
Quantity 5 kg
Assay Percent Range 99.0% min.
Beilstein 3595639
Solubility Information Soluble in water,ethanol,methanol and hydrazine.
Formula Weight 82.03
Grade ACS
Odor Nearly odorless
Sensitivity Hygroscopic
Appearance White crystalline powder
Density 1.528
Chemical Name or Material Sodium acetate, anhydrous, For ACS analysis
Show More Show Less
Hazard Category H500
EINECSNumber 204-823-8
RTECSNumber AJ4300010
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only