missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, anhydrous, 99%

Catalog No. p-7021019
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1318430 250 g
AAA131840B 1000 g
AAA131840I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1318430 Supplier Thermo Scientific Chemicals Supplier No. A1318430

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium acetate is used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, as a plating agent, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography. It is used as a base in several organic reactions in research laboratories and industries.

Solubility
Very soluble in water. Soluble in alcohol.

Notes
Hygroscopic. It is incompatible with strong oxidizing agents, halogens and it is moisture sensitive.

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 324°C
Density 1.528
pH 7.5 to 9
Assay Percent Range 99%
Linear Formula NaOOCCH3
Quantity 250 g
Beilstein 3595639
Sensitivity Hygroscopic
Merck Index 14,8571
Solubility Information Very soluble in water. Soluble in alcohol.
IUPAC Name sodium acetate
Formula Weight 82.03
Percent Purity 99%
Chemical Name or Material Sodium acetate, anhydrous
Show More Show Less
Hazard Category H500
EINECSNumber 204-823-8
RTECSNumber AJ4300010
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only