missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, ACS Reagent, Honeywell Fluka™

Catalog No. 6002199
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002199 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002199 Supplier Honeywell Chemicals Supplier No. 2412452.5KG
Only null left

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium;acetate
SMILES [Na+].CC([O-])=O

Specifications

Quantity 2.5 kg
Linear Formula CH3COONa
UN Number NONH for all modes of transport
Beilstein 3595639
Flash Point >250°C (>482°F)
Formula Weight 82.03g/mol
Percent Purity ≥99%
Grade ACS Reagent
Ignition Point 1112°F
Chemical Name or Material Sodium Acetate
EINECSNumber 204-823-8
RTECSNumber AJ4300010