missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate ACS AR, Macron Fine Chemicals™

macron

$3388.11 - $3388.11

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MK7372500
View Documents
Avantor
737212
500 g POLYSTORMOR
Case of 4 Each for $3,388.11
Only null left
 
Description

Description

Specifications

Chemical Identifiers

127-09-3
82.03
VMHLLURERBWHNL-UHFFFAOYSA-M
517045
sodium acetate
C2H3NaO2
MFCD00012459
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
CHEBI:32954
[Na+].CC([O-])=O

Specifications

127-09-3
C2H3NaO2
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
[Na+].CC([O-])=O
82.03
CHEBI:32954
ACS
Sodium Acetate, Anhydrous
500 g
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate
517045
82.03
POLYSTORMOR
Videos
SDS
Documents

Documents