Learn More
Sodium acetate, 3M aq. soln., pH 7.0, RNAse free, Thermo Scientific Chemicals
$231.26 - $231.26
Chemical Identifiers
| CAS | 127-09-3 |
|---|---|
| Molecular Formula | C2H3NaO2 |
| Molecular Weight (g/mol) | 82.03 |
| MDL Number | MFCD00012459 |
| InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
| Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
| PubChem CID | 517045 |
| ChEBI | CHEBI:32954 |
| SMILES | [Na+].CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ61934AK
|
Thermo Scientific Chemicals
J61934AK |
250 mL |
Each for $231.26
|
|
|||||
Description
Sodium acetate buffer solution pH, 7.0 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium acetate buffer solution pH, 7.0 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.
Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, halogens. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong oxidizing agent. .
Chemical Identifiers
| 127-09-3 | |
| 82.03 | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| 517045 | |
| [Na+].CC([O-])=O |
| C2H3NaO2 | |
| MFCD00012459 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| CHEBI:32954 |
Specifications
| 127-09-3 | |
| Liquid | |
| C2H3NaO2 | |
| MFCD00012459 | |
| Soluble in water (246g/L) at 20°C,and ethanol. | |
| [Na+].CC([O-])=O | |
| 82.03 | |
| CHEBI:32954 | |
| 3M aq. soln. |
| 7.0 | |
| 250 mL | |
| NaOOCCH3 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| sodium;acetate | |
| 517045 | |
| 82.03 | |
| Sodium acetate, RNAse free |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only