Learn More
Sodium acetate, 3M aq. soln., pH 5.2, RNAse free
Description
Sodium acetate buffer solution pH, 5.2 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium acetate buffer solution pH, 5.2 is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as Chip-on-Chip assay and in vitro enzyme inhibition assay.
Solubility
Soluble in water (246 g/L) at 20°C, and ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, halogens. Moisture sensitive. Stable under recommended storage conditions. Keep away from strong oxidizing agent. .
Specifications
Specifications
| pH | 5.2 |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Linear Formula | NaOOCCH3 |
| Solubility Information | Soluble aq. soln. (246 g/L) (at 20°C,and ethanol.) |
| Formula Weight | 82.03 |
| Concentration or Composition (by Analyte or Components) | 3 M aq. soln. |
| Odor | Odorless |
| Chemical Name or Material | Sodium acetate, RNAse free |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only