Learn More
Sodium acetate, 1M aq. soln., pH 4.5, RNAse free
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
$233.01 - $233.01
Chemical Identifiers
| CAS | 127-09-3 |
|---|---|
| Molecular Formula | C2H3NaO2 |
| Molecular Weight (g/mol) | 82.03 |
| MDL Number | MFCD00012459 |
| InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
| Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
| PubChem CID | 517045 |
| ChEBI | CHEBI:32954 |
| SMILES | [Na+].CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ63669AK
|
Thermo Scientific Chemicals
J63669AK |
250 mL |
Each for $233.01
|
|
|||||
Description
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. And also used in the purification and precipitation of nucleic acids, in protein crystallization, Staining of gels in protein gel electrophoresis and HPLC. Mordant dyeing; DNA microarray studies of E. coli response. Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. As a substrate of acetokinase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. And also used in the purification and precipitation of nucleic acids, in protein crystallization, Staining of gels in protein gel electrophoresis and HPLC. Mordant dyeing; DNA microarray studies of E. coli response. Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. As a substrate of acetokinase.
Solubility
Fully miscible.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 127-09-3 | |
| 82.03 | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| 517045 | |
| [Na+].CC([O-])=O |
| C2H3NaO2 | |
| MFCD00012459 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| CHEBI:32954 |
Specifications
| 127-09-3 | |
| Liquid | |
| C2H3NaO2 | |
| MFCD00012459 | |
| Fully miscible. | |
| [Na+].CC([O-])=O | |
| 82.03 | |
| CHEBI:32954 | |
| 1M aq. soln. | |
| Sodium acetate, RNAse free |
| 4.5 | |
| 250 mL | |
| NaOOCCH3 | |
| sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
| VMHLLURERBWHNL-UHFFFAOYSA-M | |
| sodium acetate | |
| 517045 | |
| 82.03 | |
| Odorless |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only