missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-hydroxybenzoate, 99+%

Catalog No. AAH5659030
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5659030 250 g
AAH5659022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5659030 Supplier Thermo Scientific Chemicals Supplier No. H5659030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 114-63-6 | C7H5NaO3 | 160.10 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 114-63-6
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.10
MDL Number MFCD00016530
InChI Key ZLVSYODPTJZFMK-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzoate, sodium p-hydroxybenzoate, 4-hydroxybenzoic acid sodium salt, sodium paraben, benzoic acid, 4-hydroxy-, monosodium salt, p-hydroxybenzoic acid sodium salt, monosodium p-hydroxybenzoate, monosodium 4-hydroxybenzoate, benzoic acid, p-hydroxy-, sodium salt, benzoic acid, 4-hydroxy-, sodium salt 1:1
PubChem CID 16219477
SMILES [Na+].OC1=CC=C(C=C1)C([O-])=O

Specifications

Quantity 250 g
Beilstein 4163553
Solubility Information Soluble in water.
IUPAC Name sodium 4-hydroxybenzoate
Formula Weight 160.1
Percent Purity ≥99%
Chemical Name or Material Sodium 4-hydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-051-1
RTECSNumber DH2900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only