missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Hydroxybenzenesulfonate Dihydrate 98.0+%, TCI America™

Catalog No. S0824500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0824500G 500 g
S082425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0824500G Supplier TCI America Supplier No. S0824500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10580-19-5
Molecular Formula C6H5O4S
Molecular Weight (g/mol) 173.16
MDL Number MFCD00044734,MFCD00150724
InChI Key FEPBITJSIHRMRT-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate
PubChem CID 23666329
IUPAC Name 4-hydroxybenzene-1-sulfonate
SMILES OC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 196.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-Hydroxybenzenesulfonate Dihydrate
EINECSNumber (3)-1958
TSCA Yes