missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-hydroxybenzenesulfonate dihydrate, 97%

Catalog No. AAA1480822
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1480822 100 g
AAA1480836 500 g
AAA148080E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1480822 Supplier Thermo Scientific Chemicals Supplier No. A1480822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10580-19-5 | C6H5O4S | 173.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10580-19-5
Molecular Formula C6H5O4S
Molecular Weight (g/mol) 173.16
MDL Number MFCD00044734,MFCD00150724
InChI Key FEPBITJSIHRMRT-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate
PubChem CID 23666329
IUPAC Name sodium;4-hydroxybenzenesulfonate;dihydrate
SMILES OC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 100 g
Beilstein 4040891
Formula Weight 232.20 (196.17 Anhydrous)
Percent Purity 97%
Chemical Name or Material Sodium 4-hydroxybenzenesulfonate dihydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-550-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only