missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-ethynylbenzoate, 97%

Catalog No. AAL1929206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1929206 5 g
AAL1929203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1929206 Supplier Thermo Scientific Chemicals Supplier No. L1929206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 144693-65-2 | C9H5NaO2 | 168.127 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144693-65-2
Molecular Formula C9H5NaO2
Molecular Weight (g/mol) 168.127
MDL Number MFCD03426026
InChI Key VXBLAFDQGACQAR-UHFFFAOYSA-M
Synonym sodium 4-ethynylbenzoate, 4-ethynylbenzoic acid sodium salt, 4-ethynyl-benzoic acid sodium salt, benzoic acid, 4-ethynyl-, sodium salt, benzoic acid,4-ethynyl-, sodium salt 1:1, 4-ethynyl benzoic acid sodium salt, sodium p-ethynylbenzoate, acmc-20egl3, 4-ethynylbenzoic acid sodium, 4-ethynylbenzoic-acid sodium salt
PubChem CID 23720426
IUPAC Name sodium;4-ethynylbenzoate
SMILES C#CC1=CC=C(C=C1)C(=O)[O-].[Na+]

Specifications

Quantity 5 g
Beilstein 5836538
Formula Weight 168.13
Percent Purity 97%
Chemical Name or Material Sodium 4-ethynylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only