missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-(2-Bromoethyl)benzenesulfonate 98.0+%, TCI America™

Catalog No. p-7135906
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
S04845G 5 g
S048425G 25 g
2 options

Catalog No. S04845G

Supplier: TCI America S04845G

Only null left

Chemical Identifiers

CAS 65036-65-9
Molecular Formula C8H8BrNaO3S
Molecular Weight (g/mol) 287.103
MDL Number MFCD06797167
InChI Key ZFGDZMNLHTUEAV-UHFFFAOYSA-M
Synonym 4-(2-Bromoethyl)benzenesulfonic Acid Sodium Salt
PubChem CID 23677320
IUPAC Name sodium;4-(2-bromoethyl)benzenesulfonate
SMILES C1=CC(=CC=C1CCBr)S(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 287.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 4-(2-Bromoethyl)benzenesulfonate