missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-Morpholinopropanesulfonate 98.0+%, TCI America™

Supplier:  TCI America M0755250G

 View more versions of this product

Catalog No. M0755250G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium 3-Morpholinopropanesulfonate
White
MFCD00064350
mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt
C1COCCN1CCCS(=O)(=O)[O-].[Na+]
231.242
231.24
Crystalline Powder
71119-22-7
C7H14NNaO4S
250 g
MWEMXEWFLIDTSJ-UHFFFAOYSA-M
sodium;3-morpholin-4-ylpropane-1-sulfonate
3859613
≥98.0% (T)

Chemical Identifiers

71119-22-7
231.242
MWEMXEWFLIDTSJ-UHFFFAOYSA-M
3859613
C1COCCN1CCCS(=O)(=O)[O-].[Na+]
C7H14NNaO4S
MFCD00064350
mops sodium salt, sodium 3-morpholinopropanesulfonate, 4-morpholinepropanesulfonic acid, sodium salt, sodium 3-morpholin-4-ylpropane-1-sulfonate, 3-n-morpholino propanesulfonic acid sodium salt, 3-4-morpholino propanesulfonic acid sodium salt, 4-morpholinepropanesulfonic acid, sodium salt 1:1, 4-morpholinepropanesulfonic acid sodium salt, sodium 3-morpholin-4-yl propane-1-sulfonate, 3-morpholin-4-ylpropanesulfonic acid, sodium salt
sodium;3-morpholin-4-ylpropane-1-sulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions