missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-Mercapto-1-propanesulfonate 85.0+%, TCI America™

Catalog No. p-7132585
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M118925G 25 g
M1189100G 100 g
M1189500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M118925G Supplier TCI America Supplier No. M118925G

Chemical Identifiers

CAS 17636-10-1
Molecular Formula C3H7NaO3S2
Molecular Weight (g/mol) 178.196
MDL Number MFCD00043148
InChI Key FRTIVUOKBXDGPD-UHFFFAOYSA-M
Synonym 3-Mercapto-1-propanesulfonic Acid Sodium Salt
PubChem CID 23672317
IUPAC Name sodium;3-sulfanylpropane-1-sulfonate
SMILES C(CS)CS(=O)(=O)[O-].[Na+]

Specifications

Melting Point 220°C
Color White
Quantity 25 g
Formula Weight 178.20
Percent Purity ≥85.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 3-Mercapto-1-propanesulfonate
EINECSNumber (2)-1597
RTECSNumber TZ6655600
TSCA Yes