missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-hydroxybutyrate, 98%

Catalog No. p-7081551
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1161306 5 g
AAA1161314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1161306 Supplier Thermo Scientific Chemicals Supplier No. A1161306
Only null left

CAS: 150-83-4 | C4H7NaO3 | 126.087 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 150-83-4
Molecular Formula C4H7NaO3
Molecular Weight (g/mol) 126.087
MDL Number MFCD00016716
InChI Key NBPUSGBJDWCHKC-UHFFFAOYSA-M
Synonym sodium 3-hydroxybutyrate, sodium 3-hydroxybutanoate, dl-3-hydroxybutyric acid sodium salt, sodium beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, monosodium salt, butanoic acid,3-hydroxy-, sodium salt 1:1, dl-3-hydroxybutyric acid, sodium salt, 3-hydroxybutyric acid sodium salt, sodium 3-oxidanylbutanoate, dl-beta-hydroxybutyric acid sodium salt
PubChem CID 23676771
IUPAC Name sodium;3-hydroxybutanoate
SMILES CC(CC(=O)[O-])O.[Na+]

Specifications

Melting Point 161°C to 167°C
Quantity 5 g
Beilstein 4824866
Sensitivity Hygroscopic
Formula Weight 126.09
Percent Purity 98%
Chemical Name or Material Sodium 3-hydroxybutyrate
EINECSNumber 205-774-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only