missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich (+/-)-Sodium 3-hydroxybutyrate

Catalog No. 5496550GF
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5496550GF 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5496550GF Supplier Sigma Aldrich Supplier No. 5496550GF

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+/-)-Sodium 3-hydroxybutyrate

Specifications

CAS 150-83-4
Melting Point 167°C to 170°C
Molecular Formula C4H7NaO3
Linear Formula CH3CH(OH)CH2COONa
MDL Number MFCD00016716
Quantity 50 g
Synonym (+/-)-3-Hydroxybutyric acid sodium salt
Molecular Weight (g/mol) 126.09
Percent Purity ≥99.0% (NT)