missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 2-Morpholinoethanesulfonate 97.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Formula Weight | 217.21 |
| Percent Purity | ≥97.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sodium 2-Morpholinoethanesulfonate |
Safety and Handling
| TSCA | Yes |