missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Sodium 2-ethylhexyl sulfate
Description
- Please note that sigma-aldrich provides this product to early discovery researchers as part of a collection of unique chemicals
- Sigma-aldrich does not collect analytical data for this product
- Buyer assumes responsibility to confirm product identity and/or purity
- All sales are final.notwithstanding any contrary provision contained in sigma-aldrich's standard terms and conditions of sale or an agreement between sigma-aldrich and buyer, sigma-aldrich sells this product as-is and makes no representation or warranty whatsoever with respect to this product, including any (a) warranty of merchantability; (b) warranty of fitness for a particular purpose; or (c) warranty against infringement of intellectual property rights of a third party; whether arising by law, course of dealing, course of performance, usage of trade or otherwise.product of otava
Specifications
Specifications
| CAS | 126-92-1 |
| Density | 1.12 g/mL (at 20°C (literature)) |
| Molecular Formula | C8H17NaO4S |
| Linear Formula | C8H17NaO4S |
| MDL Number | MFCD00042047 |
| Quantity | 1 g |
| Synonym | 2-Ethylhexyl sulfate sodium salt; 2-Ethylhexyl sulfate sodium salt |
| Molecular Weight (g/mol) | 232.27 |
| Percent Purity | 4.95%Na basis (ICP) |