missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 2-Ethylhexanoate 98.0+%, TCI America™
$32.72 - $111.66
Chemical Identifiers
| CAS | 19766-89-3 |
|---|---|
| Molecular Formula | C8H15NaO2 |
| Molecular Weight (g/mol) | 166.20 |
| MDL Number | MFCD00014007 |
| InChI Key | VYPDUQYOLCLEGS-UHFFFAOYNA-M |
| Synonym | sodium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, sodium salt, 2-ethylhexanoic acid sodium salt, sodium-2-ethylcaproate, 2-ethylcaproic acid sodium salt, hexanoic acid, 2-ethyl-, sodium salt 1:1, sodium 2ethylhexanoate, sodium 2-ethylcaproate, sodium 2-ethyl-hexanoate |
| PubChem CID | 23672331 |
| IUPAC Name | sodium 2-ethylhexanoate |
| SMILES | [Na+].CCCCC(CC)C([O-])=O |
Chemical Identifiers
| 19766-89-3 | |
| 166.20 | |
| VYPDUQYOLCLEGS-UHFFFAOYNA-M | |
| 23672331 | |
| [Na+].CCCCC(CC)C([O-])=O |
| C8H15NaO2 | |
| MFCD00014007 | |
| sodium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, sodium salt, 2-ethylhexanoic acid sodium salt, sodium-2-ethylcaproate, 2-ethylcaproic acid sodium salt, hexanoic acid, 2-ethyl-, sodium salt 1:1, sodium 2ethylhexanoate, sodium 2-ethylcaproate, sodium 2-ethyl-hexanoate | |
| sodium 2-ethylhexanoate |
Specifications
| 19766-89-3 | |
| C8H15NaO2 | |
| 25 g | |
| VYPDUQYOLCLEGS-UHFFFAOYNA-M | |
| sodium 2-ethylhexanoate | |
| 23672331 | |
| ≥98.0% (T) | |
| Sodium 2-Ethylhexanoate |
| White | |
| MFCD00014007 | |
| sodium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, sodium salt, 2-ethylhexanoic acid sodium salt, sodium-2-ethylcaproate, 2-ethylcaproic acid sodium salt, hexanoic acid, 2-ethyl-, sodium salt 1:1, sodium 2ethylhexanoate, sodium 2-ethylcaproate, sodium 2-ethyl-hexanoate | |
| [Na+].CCCCC(CC)C([O-])=O | |
| 166.20 | |
| 166.20 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0611
RTECSNumber : MO7855000
TSCA : Yes