missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Ethylhexanoate 98.0+%, TCI America™

Catalog No. S086225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S086225G 25 g
S0862250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S086225G Supplier TCI America Supplier No. S086225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19766-89-3
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.20
MDL Number MFCD00014007
InChI Key VYPDUQYOLCLEGS-UHFFFAOYNA-M
Synonym sodium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, sodium salt, 2-ethylhexanoic acid sodium salt, sodium-2-ethylcaproate, 2-ethylcaproic acid sodium salt, hexanoic acid, 2-ethyl-, sodium salt 1:1, sodium 2ethylhexanoate, sodium 2-ethylcaproate, sodium 2-ethyl-hexanoate
PubChem CID 23672331
IUPAC Name sodium 2-ethylhexanoate
SMILES [Na+].CCCCC(CC)C([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 166.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Ethylhexanoate
EINECSNumber (2)-0611
RTECSNumber MO7855000
TSCA Yes