missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-ethylhexanoate, 97%

Catalog No. p-7027899
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2496030 250 g
AAB249600B 1000 g
AAB249600I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2496030 Supplier Thermo Scientific Chemicals Supplier No. B2496030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19766-89-3 | C8H15NaO2 | 166.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium 2-ethylhexanoate is useful in acylation of amines. It is an active ingredient in wood preservative agent. It is mainly used for synthetic 2-ethylhexanoic acid and calcium, magnesium salts. It is also used in salt forming agent of pharmaceutical, dry catalytic agent of the paint, polymer stabilizers, crosslinking agent, thickening agent of oil, and energy-saving additive of fuel.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 19766-89-3
Molecular Formula C8H15NaO2
Molecular Weight (g/mol) 166.20
MDL Number MFCD00014007
InChI Key VYPDUQYOLCLEGS-UHFFFAOYNA-M
Synonym sodium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, sodium salt, 2-ethylhexanoic acid sodium salt, sodium-2-ethylcaproate, 2-ethylcaproic acid sodium salt, hexanoic acid, 2-ethyl-, sodium salt 1:1, sodium 2ethylhexanoate, sodium 2-ethylcaproate, sodium 2-ethyl-hexanoate
PubChem CID 23672331
IUPAC Name sodium;2-ethylhexanoate
SMILES [Na+].CCCCC(CC)C([O-])=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 250 g
Solubility Information Soluble in water.
Formula Weight 166.2
Percent Purity 97%
Chemical Name or Material Sodium 2-ethylhexanoate
Hazard Category H361
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 243-283-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only