missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Bromoethanesulfonate 97.0+%, TCI America™

Catalog No. B058925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B058925G 25 g
B0589500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B058925G Supplier TCI America Supplier No. B058925G

Chemical Identifiers

CAS 4263-52-9
Molecular Formula C2H4BrNaO3S
Molecular Weight (g/mol) 211.005
MDL Number MFCD00007530
InChI Key HNFOAHXBHLWKNF-UHFFFAOYSA-M
Synonym sodium 2-bromoethanesulfonate, 2-bromoethanesulfonic acid sodium salt, sodium 2-bromoethanesulphonate, 2-bromoethanesulfonic acid, sodium salt, sodium bromoethylsulfonate, ethanesulfonic acid, 2-bromo-, sodium salt, ethanesulfonic acid, 2-bromo-, sodium salt 1:1, sodium 2-bromoethane sulfonate, 2-bromoethanesulfonic acid sodium, natrium-2-bromethansulfonat
PubChem CID 23666797
IUPAC Name sodium;2-bromoethanesulfonate
SMILES C(CBr)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 286°C
Color White-Yellow
Quantity 25 g
Formula Weight 211.00
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Bromoethanesulfonate
EINECSNumber (2)-1587
TSCA Yes